siRNA Details
Gene Symbol: Prr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126375.1 | NP_001119847.2 | 2 | 1271 | ||
XM_006244338.2 | XP_006244400.1 | 3 | 1252 | |||
GenBank mRNA | BC161872.1 | - | 2 | 1271 | 7112899 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
