siRNA Details
Gene Symbol: Snrpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109399.2 | NP_001102869.1 | 2 | 182 | ||
GenBank mRNA | BC168781.1 | - | 2 | 182 | 6921356 | |
FQ222619.1 | - | 2 | 165 | |||
FQ228670.1 | - | 2 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
