siRNA Details
Gene Symbol: Tceal7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109401.1 | NP_001102871.1 | 3 | 297 | ||
XM_006257279.2 | XP_006257341.1 | 3 | 334 | |||
XM_006257280.2 | XP_006257342.1 | 3 | 313 | |||
XM_008773437.1 | XP_008771659.1 | 3 | 329 | |||
GenBank mRNA | FQ214942.1 | - | 3 | 302 | ||
FQ215364.1 | - | 3 | 286 | |||
FQ215415.1 | - | 3 | 321 | |||
FQ216235.1 | - | 3 | 307 | |||
FQ216331.1 | - | 3 | 302 | |||
FQ217308.1 | - | 3 | 307 | |||
FQ224746.1 | - | 3 | 302 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001127169, BY775878, AK003800.
Gene Ontology Categories:
