siRNA Details
Gene Symbol: Tceal7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109401.1 | NP_001102871.1 | 3 | 352 | ||
XM_006257279.2 | XP_006257341.1 | 3 | 389 | |||
XM_006257280.2 | XP_006257342.1 | 3 | 368 | |||
XM_008773437.1 | XP_008771659.1 | 3 | 384 | |||
GenBank mRNA | FQ215364.1 | - | 3 | 341 | ||
FQ215415.1 | - | 3 | 376 | |||
FQ216235.1 | - | 3 | 362 | |||
FQ216331.1 | - | 3 | 357 | |||
FQ217308.1 | - | 3 | 362 | |||
FQ224746.1 | - | 3 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
