siRNA Details
Gene Symbol: Clec12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134716.1 | NP_001128188.1 | 245 | |||
GenBank mRNA | BC168186.1 | - | 245 | 7100019 | ||
GU357484.1 | - | 44 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Clec12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134716.1 | NP_001128188.1 | 245 | |||
GenBank mRNA | BC168186.1 | - | 245 | 7100019 | ||
GU357484.1 | - | 44 |