siRNA Details
Gene Symbol: Wbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127502.1 | NP_001120974.1 | 3 | 501 | ||
XM_006257281.2 | XP_006257343.1 | 3 | 540 | |||
XM_008773438.1 | XP_008771660.1 | 3 | 521 | |||
GenBank mRNA | BC166897.1 | - | 3 | 501 | 6921863 | |
FQ215245.1 | - | 3 | 429 | |||
FQ220910.1 | - | 3 | 483 | |||
FQ232216.1 | - | 3 | 475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
