siRNA Details
Gene Symbol: Mark4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191071.1 | NP_001178000.1 | 4 | 355 | ||
XM_006228472.2 | XP_006228534.1 | 4 | 395 | |||
XM_008758981.1 | XP_008757203.1 | 4 | 395 |
