siRNA Details
Gene Symbol: Dazl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109414.1 | NP_001102884.1 | 3 | 416 | ||
XM_006244477.2 | XP_006244539.1 | 3 | 269 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001277863, NM_010021, X95724, BC099940, U46694, and others.
Gene Ontology Categories:
