siRNA Details
Gene Symbol: Fam64a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113781.2 | NP_001107253.2 | 2 | 96 | ||
XM_006246759.2 | XP_006246821.1 | 2 | 206 | |||
XM_006246760.2 | XP_006246822.1 | 2 | 182 | |||
GenBank mRNA | BC158692.1 | - | 2 | 96 | 9022735 |
Gene Symbol: LOC100912261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766825.1 | XP_008765047.1 | 1 | 57 |

More Information
Other Species Targeted

