siRNA Details
Gene Symbol: LOC680590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CK597145.1 | - | 3, 4 | 160 | ||
CK652853.1 | - | 3, 4 | 152 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC680590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CK597145.1 | - | 3, 4 | 160 | ||
CK652853.1 | - | 3, 4 | 152 |