siRNA Details
Gene Symbol: Dph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134850.1 | NP_001128322.1 | 3 | 619 | ||
XM_006252668.1 | XP_006252730.1 | 2 | 492 | |||
GenBank mRNA | BC168674.1 | - | 2 | 2347 | 7939231 |
