siRNA Details
Gene Symbol: Fcrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232673.2 | XP_006232735.1 | 10 | 2441 | ||
XM_006232674.2 | XP_006232736.1 | 9 | 2420 | |||
GenBank mRNA | FQ232638.1 | - | 10 | 1371 |
