siRNA Details
Gene Symbol: Casp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767527.1 | XP_008765749.1 | 4 | 1856 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Casp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767527.1 | XP_008765749.1 | 4 | 1856 |