siRNA Details
Gene Symbol: Ocel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106065.1 | NP_001099535.1 | 4 | 996 | ||
XM_006252856.2 | XP_006252918.1 | 3 | 1626 | |||
XM_008771080.1 | XP_008769302.1 | 3 | 1345 | |||
GenBank mRNA | BC169113.1 | - | 3 | 1088 | 7445881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
