siRNA Details
Gene Symbol: Tmem45al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751037.3 | XP_003751085.1 | 3 | 806 | ||
XM_006248218.2 | XP_006248280.1 | 3 | 809 | |||
XM_008768662.1 | XP_008766884.1 | 3 | 810 | |||
XM_008768663.1 | XP_008766885.1 | 3 | 806 |
Gene Symbol: Tmem45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191074.1 | NP_001178003.1 | 3 | 714 |
