siRNA Details
Gene Symbol: Fam169b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229362.2 | XP_006229424.1 | 5 | 561 | ||
XM_006229363.2 | XP_006229425.1 | 6 | 738 | |||
XM_008759577.1 | XP_008757799.1 | 6 | 654 | |||
XM_008759578.1 | XP_008757800.1 | 6 | 687 | |||
XM_008759579.1 | XP_008757801.1 | 6 | 650 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
