siRNA Details
Gene Symbol: Bmp8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109432.1 | NP_001102902.1 | 7 | 1796 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bmp8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109432.1 | NP_001102902.1 | 7 | 1796 |