siRNA Details
Gene Symbol: Dnajc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109024.2 | NP_001102494.1 | 1 | 464 | ||
GenBank mRNA | BC166911.1 | - | 1 | 464 | 6890506 | |
FQ214485.1 | - | 1 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
