siRNA Details
Gene Symbol: LOC302022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127530.1 | NP_001121002.1 | 1277 | |||
GenBank mRNA | BC162000.1 | - | 1277 | 9036094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
