siRNA Details
Gene Symbol: LOC681077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228551.2 | XP_006228613.1 | 715 | |||
XM_008759071.1 | XP_008757293.1 | 448 | ||||
XM_008759072.1 | XP_008757294.1 | 4 | 716 | |||
XM_008759073.1 | XP_008757295.1 | 787 | ||||
XM_008759074.1 | XP_008757296.1 | 3 | 608 |
Gene Symbol: LOC100910611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759098.1 | XP_008757320.1 | 3 | 448 |
Gene Symbol: LOC100910953 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228539.2 | XP_006228601.1 | 4 | 715 | ||
XM_008759066.1 | XP_008757288.1 | 3 | 448 | |||
XM_008759067.1 | XP_008757289.1 | 4 | 787 |
