siRNA Details
Gene Symbol: Pigf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239725.2 | XP_006239787.1 | 2 | 259 | ||
XM_008764472.1 | XP_008762694.1 | 2 | 298 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pigf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239725.2 | XP_006239787.1 | 2 | 259 | ||
XM_008764472.1 | XP_008762694.1 | 2 | 298 |