siRNA Details
Gene Symbol: Cks1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135749.1 | NP_001129221.1 | 2, 3 | 231 | ||
GenBank mRNA | BC167077.1 | - | 2, 3 | 231 | 9023967 | |
BC168780.1 | - | 2, 3 | 263 | 6918892 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_016904, BC076579, AK011314, AK010414, AK011782, and others.
Gene Ontology Categories:
