siRNA Details
Gene Symbol: Cks1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135749.1 | NP_001129221.1 | 2 | 172 | ||
GenBank mRNA | BC167077.1 | - | 2 | 172 | 9023967 | |
BC168780.1 | - | 2 | 204 | 6918892 |
