siRNA Details
Gene Symbol: Kcne5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101003.1 | NP_001094473.1 | 1 | 261 | ||
GenBank mRNA | EF535526.1 | - | 1 | 261 |

More Information
Other Species Targeted

Gene Ontology Categories:
