siRNA Details
Gene Symbol: Cox14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001060794.5 | XP_001060794.2 | 2 | 257 | ||
GenBank mRNA | BC084716.1 | - | 2 | 176 | 7314502 |

More Information
Other Species Targeted

Gene Ontology Categories:
