siRNA Details
Gene Symbol: LOC102549803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236818.2 | XP_006236880.1 | 2 | 2243 | ||
XM_008763111.1 | XP_008761333.1 | 3 | 1823 | |||
XM_008763112.1 | XP_008761334.1 | 3 | 2050 | |||
XM_008763113.1 | XP_008761335.1 | 3 | 1426 |
Gene Symbol: LOC102556148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236802.2 | XP_006236864.1 | 1426 | |||
XM_008763098.1 | XP_008761320.1 | 2 | 3460 | |||
XM_008763099.1 | XP_008761321.1 | 3 | 3040 | |||
XM_008763100.1 | XP_008761322.1 | 2050 |
