siRNA Details
Gene Symbol: ENTPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128930.2 | NP_001122402.1 | 4 | 512 | ||
NM_004901.4 | NP_004892.1 | 4 | 512 | |||
GenBank mRNA | AB002390.1 | - | 2 | 66 | ||
AF016032.1 | - | 4 | 441 | |||
AJ131358.1 | - | 4 | 409 | |||
AK300464.1 | - | 4 | 420 | |||
BC034477.2 | - | 4 | 476 | 4824559 | ||
CN313716.1 | - | 2 | 68 | |||
DC341200.1 | - | 4 | 512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
