siRNA Details
Gene Symbol: Cxxc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007628.2 | NP_001007629.1 | 2 | 1192 | ||
GenBank mRNA | BC083622.1 | - | 2 | 1192 | 7134100 | |
DV723039.1 | - | 1 | 75 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133687, AK005364, BC016207, BC089314, AK015150, and others.
Gene Ontology Categories:
