siRNA Details
Gene Symbol: Letmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122781.3 | NP_001116253.3 | 2 | 223 | ||
XM_006242354.2 | XP_006242416.1 | 2 | 286 | |||
GenBank mRNA | BC159426.1 | - | 2 | 204 | 9034024 | |
CB778986.1 | - | 2 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
