siRNA Details
Gene Symbol: Rnf133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044278.1 | NP_001037743.1 | 1 | 656 | ||
XM_006236144.2 | XP_006236206.1 | 2 | 2568 | |||
GenBank mRNA | BC079249.1 | - | 1 | 656 | 7133029 |

More Information
Other Species Targeted

Gene Ontology Categories:
