siRNA Details
Gene Symbol: Rps27l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276477.1 | NP_001263406.1 | 3 | 355 | ||
GenBank mRNA | CB712651.1 | - | 2 | 105 | ||
FQ109437.1 | - | 3 | 354 | |||
FQ209888.1 | - | 2 | 152 | |||
X59375.1 | - | 2 | 153 |

More Information
Other Species Targeted

Gene Ontology Categories:
