siRNA Details
Gene Symbol: Spcs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191073.1 | NP_001178002.1 | 3 | 278 | ||
GenBank mRNA | AY724480.1 | - | 397 |

More Information
Other Species Targeted

Gene Ontology Categories:
