siRNA Details
Gene Symbol: Rnf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109444.2 | NP_001102914.1 | 314 | |||
GenBank mRNA | BC081881.1 | - | 314 | 7108181 | ||
FQ224754.1 | - | 482 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109444.2 | NP_001102914.1 | 314 | |||
GenBank mRNA | BC081881.1 | - | 314 | 7108181 | ||
FQ224754.1 | - | 482 |