siRNA Details
Gene Symbol: Pdzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106362.2 | NP_001099832.2 | 1 | 257 | ||
XM_006231449.2 | XP_006231511.1 | 2 | 863 | |||
XM_008760406.1 | XP_008758628.1 | 2 | 863 | |||
XM_008760407.1 | XP_008758629.1 | 2 | 863 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
