siRNA Details
Gene Symbol: LOC103693049 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766089.1 | XP_008764311.1 | 1 | 409 |
Gene Symbol: LOC100911359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766100.1 | XP_008764322.1 | 2 | 592 |
