siRNA Details
Gene Symbol: Tmem41a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100805.1 | NP_001094275.1 | 4, 5 | 639 | ||
GenBank mRNA | AY313923.1 | - | 3, 4 | 415 | ||
BC161997.1 | - | 4, 5 | 639 | 9035693 |
