siRNA Details
Gene Symbol: Fam208a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001055921.4 | XP_001055921.3 | 3, 4 | 1728 | ||
XM_003751541.3 | XP_003751589.1 | 3, 4 | 1728 | |||
XM_006252675.1 | XP_006252737.1 | 3, 4 | 1728 |

More Information
Other Species Targeted

Gene Ontology Categories:
