siRNA Details
Gene Symbol: Ndufb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037338.1 | NP_001032415.1 | 2 | 231 | ||
GenBank mRNA | CB732095.1 | - | 2 | 231 | ||
DQ255901.1 | - | 2 | 84 | |||
FQ217067.1 | - | 2 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
