siRNA Details
Gene Symbol: Msrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044285.2 | NP_001037750.2 | 2 | 253 | ||
GenBank mRNA | BC094309.1 | - | 2 | 243 | 7316479 | |
CB812976.1 | - | 2 | 253 | |||
FQ220312.1 | - | 2 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
