siRNA Details
Gene Symbol: LOC691670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109646.1 | NP_001103116.1 | 2 | 137 | ||
XM_008770702.1 | XP_008768924.1 | 2 | 190 | |||
XM_008770703.1 | XP_008768925.1 | 2 | 190 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
