siRNA Details
Gene Symbol: Pomt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047114.1 | NP_001040579.1 | 7 | 1364 | ||
GenBank mRNA | AB246667.1 | - | 7 | 1364 |

More Information
Other Species Targeted

Gene Ontology Categories:
