siRNA Details
Gene Symbol: Slc25a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044280.2 | NP_001037745.2 | 8, 9 | 1022 | ||
GenBank mRNA | BC094517.1 | - | 8, 9 | 967 | 7314749 |

More Information
Other Species Targeted

Gene Ontology Categories:
