siRNA Details
Gene Symbol: Slc25a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044280.2 | NP_001037745.2 | 9 | 1035 | ||
GenBank mRNA | BC094517.1 | - | 9 | 980 | 7314749 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
