siRNA Details
Gene Symbol: Samd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127609.1 | NP_001121081.1 | 1 | 1348 | ||
XM_006240398.2 | XP_006240460.1 | 1 | 1156 | |||
GenBank mRNA | BC161884.1 | - | 1 | 1348 | 7131792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
