siRNA Details
Gene Symbol: Noxred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750205.3 | XP_003750253.1 | 7 | 1353 | ||
XM_006240401.2 | XP_006240463.1 | 7 | 1169 | |||
XM_006240402.2 | XP_006240464.1 | 7 | 1366 | |||
XM_006240403.2 | XP_006240465.1 | 7 | 1499 | |||
XM_008764893.1 | XP_008763115.1 | 8 | 1250 | |||
XM_008764894.1 | XP_008763116.1 | 8 | 1464 |
