siRNA Details
Gene Symbol: Mpnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085406.1 | NP_001078875.1 | 11 | 1288 | ||
XM_008766773.1 | XP_008764995.1 | 12 | 1576 | |||
GenBank mRNA | BC101893.1 | - | 10 | 1116 | ||
BC158552.1 | - | 11 | 1288 | 9098910 | ||
FQ226713.1 | - | 3 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
