siRNA Details
Gene Symbol: Smim12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276483.1 | NP_001263412.1 | 2 | 180 | ||
XM_006238907.2 | XP_006238969.1 | 3 | 329 | |||
GenBank mRNA | BP485002.1 | - | 2 | 187 | ||
DV214253.1 | - | 2 | 172 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_030252, BY237825, AK141221, AK075968, BC147068, and others.
