siRNA Details
Gene Symbol: Dennd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126289.1 | NP_001119761.1 | 8 | 954 | ||
GenBank mRNA | BC160823.1 | - | 8 | 954 | 7380256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
