siRNA Details
Gene Symbol: Jazf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001065610.5 | XP_001065610.1 | 5 | 1634 | ||
XM_006236535.2 | XP_006236597.1 | 5 | 2422 | |||
XM_006236536.2 | XP_006236598.1 | 5 | 3540 | |||
XM_008762953.1 | XP_008761175.1 | 5 | 1761 | |||
XM_008762954.1 | XP_008761176.1 | 6 | 1976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
