siRNA Details
Gene Symbol: LOC682102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134732.1 | NP_001128204.1 | 3, 4 | 1404 | ||
GenBank mRNA | BC168197.1 | - | 2, 3 | 711 | 7112960 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
